missing translation for 'onlineSavingsMsg'
Learn More

GSK461364, MedChemExpress

Product Code. 16440588
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440588

Brand: Medchem Express HY50877

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK461364 is a selective, reversible and ATP-competitive Polo-like kinase 1 (PLK1) inhibitor with a Ki value of 2.2 nM.

GSK461364 is a selective, reversible and ATP-competitive Polo-like kinase 1 (PLK1) inhibitor with a Ki value of 2.2 nM.

Specifications

Chemical Name or Material GSK461364
CAS 929095-18-1
Color Gray
Purity Grade Notes Research
Molecular Formula C27H28F3N5O2S
Quantity 5 mg
Synonym GSK461364A
Solubility Information DMSO : 50 mg/mL (91.98 mM; Need ultrasonic)
SMILES O=C(C(SC(N1C=NC2=CC=C(C=C12)CN3CCN(CC3)C)=C4)=C4O[C@@H](C5=CC=CC=C5C(F)(F)F)C)N
Molecular Weight (g/mol) 543.6
Formula Weight 543.6
Percent Purity 96.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.