missing translation for 'onlineSavingsMsg'
Learn More

GSK4112, MedChemExpress

Product Code. 16446058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446058 10 mg 10mg
16442197 50 mg 50mg
16408667 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16446058 Supplier Medchem Express Supplier No. HY14414

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK4112 is a Rev-erbα agonist with EC50 of 0.4 μM, also is a small molecule chemical probe for the cell biology of the nuclear heme receptor Rev-erbα.

GSK4112 is a Rev-erbα agonist with EC50 of 0.4 μM, also is a small molecule chemical probe for the cell biology of the nuclear heme receptor Rev-erbα.

Specifications

Chemical Name or Material GSK4112
Physical Form Solid
CAS 1216744-19-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H21ClN2O4S
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym SR6452
Solubility Information DMSO : 25 mg/mL (62.99 mM; Need ultrasonic)
SMILES ClC1=CC=C(CN(CC2=CC=C([N+]([O-])=O)S2)CC(OC(C)(C)C)=O)C=C1
Molecular Weight (g/mol) 396.89
Formula Weight 396.89
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.