missing translation for 'onlineSavingsMsg'
Learn More

GSK4028, MedChemExpress

Product Code. 16473238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473238 10 mg 10mg
16455967 5 mg 5mg
16449786 50 mg 50mg
16434506 100 mg 100mg
16422686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473238 Supplier Medchem Express Supplier No. HY101027A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK4028 is the enantiomeric negative control of GSK4027, which is a PCAF/GCN5 bromodomain chemical probe, the pIC50 of GSK4028 is 4.9 in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay.

GSK4028 is the enantiomeric negative control of GSK4027, which is a PCAF/GCN5 bromodomain chemical probe, the pIC50 of GSK4028 is 4.9 in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay.

Specifications

Chemical Name or Material GSK4028
CAS 2079886-19-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H21BrN4O
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (265.06 mM; Need ultrasonic)
SMILES BrC1=C(N[C@H]2C[C@@H](C3=CC=CC=C3)CN(C)C2)C=NN(C)C1=O
Molecular Weight (g/mol) 377.28
Formula Weight 377.28
Percent Purity 98.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.