Learn More
GSK3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22104-1-AP-20UL
 View more versions of this product
Product Code. 16881645
  • £135.00 / 20µL
Description
This antibody recognize the C-terminal of GSK3B.
Menin is a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Two variants of the shorter transcript have been identified where alternative splicing affects the CDS. Five variants where alternative splicing takes place in the 5′ UTR have also been identified.
Specifications
Specifications
GSK3B
Polyclonal
Unconjugated
GSK3B
GSK 3 beta, GSK3 beta, GSK3A B, GSK3B, GSK-3B, GSK3B 216P, gsk3beta
Rabbit
Antigen Affinity Chromatography
RUO
2932, 56637, 84027
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18266, P49841, Q9WV60
Gsk3b
GSK3B Fusion Protein Ag17320
20 μL
Primary
Human, Mouse, Rat