missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA595845
This item is not returnable.
View return policy
Description
Immunogen sequence: LPNGRDTPAL FNFTTQELSS NPPLATILIP PHARIQAAAS TPTNATAASD ANTGDRGQTN NAASASASNS T
GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
Specifications
| GSK3B | |
| Polyclonal | |
| Unconjugated | |
| GSK3B | |
| 7330414F15Rik, 8430431H08Rik, C86142, EC 2.7.11.26, FA, Factor A, fb68h05, fk80d11, glycogen synthase kinase 3, glycogen synthase kinase 3 beta, glycogen synthase kinase 3 beta L homeolog, glycogen synthase kinase 3 beta variant 1, glycogen synthase kinase 3 beta variant 2, glycogen synthase kinase 3 beta variant 3, glycogen synthase kinase 3 beta variant 4, glycogen synthase kinase 3 beta variant 5, glycogen synthase kinase 3 beta, genome duplicate a, glycogen synthase kinase-3 beta, glycogen synthase kinase-3 beta splice variant X1, glycogen synthase kinase-3 beta splice variant X2, glycogen synthase kinase-3 beta splice variant X3, GSK 3 beta, GSK 3B, gsk3, GSK-3, GSK3 beta, GSK-3 beta, GSK-3 beta GSK3B, GSK-3[b], Gsk3b, GSK-3b, gsk3b.L, gsk3ba, gsk3beta, GSK-3beta, gsk3-beta, GSK3beta isoform, intracellular kinase, pp37, serine/threonine-protein kinase GSK3B, wu:fb68h05, wu:fk80d11, XELAEV_180112401mg, xgsk-3, Xgsk-3 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2932, 56637, 84027 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
| P18266, P49841, Q9WV60 | |
| GSK3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-420 of GSK3&beta, (NP_0011396281) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction