missing translation for 'onlineSavingsMsg'
Learn More

GSK376501A, MedChemExpress

Product Code. 16486267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486267 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16486267 Supplier Medchem Express Supplier No. HY101746
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK376501A is a selective peroxisome proliferator-activated receptor gamma (PPARγ) modulator for the treatment of type 2 diabetes mellitus.

GSK376501A is a selective peroxisome proliferator-activated receptor gamma (PPARγ) modulator for the treatment of type 2 diabetes mellitus.

Specifications

Chemical Name or Material GSK376501A
CAS 1010412-80-2
Color White
Purity Grade Notes Research
Molecular Formula C32H37NO6
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (188.10 mM; Need ultrasonic)
SMILES O=C(C(N1CC2=CC(OCCOC)=CC(OCCOC)=C2)=C(C3=CC=C(C(C)(C)C)C=C3)C4=C1C=CC=C4)O
Molecular Weight (g/mol) 531.64
Formula Weight 531.64
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.