Learn More
Eurofins DiscoverX GSK3α Protein, Active
Description
Glycogen synthase kinase 3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. There exists two isoforms of GSK3, GSK3A and GSK3B, and they are strictly regulated via phosphorylation. GSK3 has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3A also has been inhibited by activation of the canonical Wnt pathway, resulting in increased catenin that drives human mesenchymal stem cells to osteoblasts.
GenBank NM_019884. The recombinant GSK3a sequence deviates from that published in NM_019884 by a single amino acid substitution, S449A. This substitution has been reported elsewhere,GenBank 2641993.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 270 mM sucrose, 0.1 mM EGTA, 150 mM NaCl, 1 mM benzamidine, 0.03% Brij-35, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 55 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥50% |
| Protein | GSK-3a, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.