missing translation for 'onlineSavingsMsg'
Learn More

GSK2983559 active metabolite, MedChemExpress

Product Code. 16487706
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487706 100 mg 100mg
16480358 5 mg 5mg
16451859 1 mg 1mg
16447898 10 mg 10mg
16413847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487706 Supplier Medchem Express Supplier No. HY19764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2983559 active metabolite is an active metabolite of GSK2983559. GSK2983559 active metabolite is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043446 A1, compound example 1.

GSK2983559 active metabolite is an active metabolite of GSK2983559. GSK2983559 active metabolite is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043446 A1, compound example 1.

Specifications

Chemical Name or Material GSK2983559 active metabolite
CAS 1423186-80-4
Color Gray
Purity Grade Notes Research
Molecular Formula C21H22N4O4S2
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (54.52 mM; Need ultrasonic)
SMILES CC(C)(C)S(C1=C(OCCO)C=C(N=CN=C2NC3=CC(N=CS4)=C4C=C3)C2=C1)(=O)=O
Molecular Weight (g/mol) 458.55
Formula Weight 458.55
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.