missing translation for 'onlineSavingsMsg'
Learn More

GSK2981278, MedChemExpress

Product Code. 16471859
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471859 1 mg 1mg
16467898 10 mg 10mg
16433847 50 mg 50mg
16407716 100 mg 100mg
16400368 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471859 Supplier Medchem Express Supplier No. HY19770
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK2981278 is a potent and selective RORγ inverse agonist. GSK2981278 inhibits activation of the il17 promoter and interferes RORγ-DNA binding.

GSK2981278 is a potent and selective RORγ inverse agonist. GSK2981278 inhibits activation of the il17 promoter and interferes RORγ-DNA binding.

Specifications

Chemical Name or Material GSK2981278
CAS 1474110-21-8
Color White
Purity Grade Notes Research
Molecular Formula C25H35NO5S
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (216.63 mM)
SMILES CC(C)CN(S(=O)(C1=CC=C(OCC2CCOCC2)C(CO)=C1)=O)C3=CC=C(CC)C=C3
Molecular Weight (g/mol) 461.61
Formula Weight 461.61
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.