missing translation for 'onlineSavingsMsg'
Learn More

GSK2801, MedChemExpress

Product Code. 16462807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462807 50 mg 50mg
16459277 5 mg 5mg
16446728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16462807 Supplier Medchem Express Supplier No. HY15658
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK2801 is a potent, selective, orally active and cell active acetyl-lysine competitive BAZ2A and BAZ2B bromodomains inhibitor with Kd values of 136 nM and 257 nM, respectively. GSK2801 shows >50-fold selectivity for BAZ2A/B over BRD4.

GSK2801 is a potent, selective, orally active and cell active acetyl-lysine competitive BAZ2A and BAZ2B bromodomains inhibitor with Kd values of 136 nM and 257 nM, respectively. GSK2801 shows >50-fold selectivity for BAZ2A/B over BRD4.

Specifications

Chemical Name or Material GSK2801
CAS 1619994-68-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H21NO4S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (134.61 mM; Need ultrasonic)
SMILES O=C(C)C1=CC(C2=C(S(=O)(C)=O)C=CC=C2)=C3N1C=CC(OCCC)=C3
Molecular Weight (g/mol) 371.45
Formula Weight 371.45
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.