missing translation for 'onlineSavingsMsg'
Learn More

GSK2795039, MedChemExpress

Product Code. 16470118
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470118

Brand: Medchem Express HY18950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2795039 is a NADPH oxidase 2 (NOX2) inhibitor with a mean pIC50 of 6 in different cell-free assays. GSK2795039 inhibits reactive oxygen species (ROS) production and NADPH consumption. GSK2795039 reduces apoptosis.

GSK2795039 is a NADPH oxidase 2 (NOX2) inhibitor with a mean pIC50 of 6 in different cell-free assays. GSK2795039 inhibits reactive oxygen species (ROS) production and NADPH consumption.

Specifications

Chemical Name or Material GSK2795039
CAS 1415925-18-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H26N6O2S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (221.95 mM; Need ultrasonic)
SMILES CC(C)N1C=C(C2=CC=C(CCN3C)C3=C2)C4=C1N=CC=C4NS(C5=NN(C)C=C5)(=O)=O
Molecular Weight (g/mol) 450.56
Formula Weight 450.56
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.