missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK269962A, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
GSK269962A (GSK 269962) is a potent ROCK inhibitor with IC50s of 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively. GSK269962A has anti-inflammatory and vasodilatory activities.
Specifications
Specifications
| Chemical Name or Material | GSK269962A |
| CAS | 850664-21-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H30N8O5 |
| Quantity | 5 mg |
| Synonym | GSK 269962 |
| Solubility Information | DMSO : ≥ 30 mg/mL (52.58 mM) |
| SMILES | CCN1C2=CC(OC3=CC=CC(NC(C4=CC=C(OCCN5CCOCC5)C=C4)=O)=C3)=NC=C2N=C1C6=NON=C6N |
| Molecular Weight (g/mol) | 570.6 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction