missing translation for 'onlineSavingsMsg'
Learn More

GSK2643943A, MedChemExpress

Product Code. 16473096
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473096 25 mg 25mg
16447187 5 mg 5mg
16440777 50 mg 50mg
16425346 100 mg 100mg
16404468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473096 Supplier Medchem Express Supplier No. HY111458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2643943A is a deubiquitinating enzyme (DUB) inhibitor targeting USP20. GSK2643943A has affinity with an IC50 of 160 nM for USP20/Ub-Rho. GSK2643943A has anti-tumor efficacy and can be used for the research of oral squamous cell carcinoma (OSCC) .

GSK2643943A is a deubiquitinating enzyme (DUB) inhibitor targeting USP20. GSK2643943A has affinity with an IC50 of 160 nM for USP20/Ub-Rho. GSK2643943A has anti-tumor efficacy and can be used for the research of oral squamous cell carcinoma (OSCC) .

Specifications

Chemical Name or Material GSK2643943A
CAS 2449301-27-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H12FN3
Quantity 25 mg
Solubility Information DMSO : ≥ 125 mg/mL (450.78 mM)
SMILES FC1=CC=CC(/C=C/C2=CC(NC(N)=C3C#N)=C3C=C2)=C1
Molecular Weight (g/mol) 277.3
Formula Weight 277.3
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.