missing translation for 'onlineSavingsMsg'
Learn More

GSK2334470, MedChemExpress

Product Code. 16456278
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456278 10 mg 10mg
16446616 100 mg 100mg
16402407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456278 Supplier Medchem Express Supplier No. HY14981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2334470 is a highly specific and potent inhibitor of PDK1 with an IC50 of 10 nM.

GSK2334470 is a highly specific and potent inhibitor of PDK1 with an IC50 of 10 nM.

Specifications

Chemical Name or Material GSK2334470
CAS 1227911-45-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H34N8O
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (108.09 mM)
SMILES O=C([C@@H]1CN(C2=NC(NC)=NC(C3=CC4=C(C=C3)C(N)=NN4)=C2)[C@H](C)CC1)NC5CCCCC5
Molecular Weight (g/mol) 462.59
Formula Weight 462.59
Percent Purity 95.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.