missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amantadine hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
Description
Amantadine hydrochloride inhibits several ion channels such as NMDA and M2, and also inhibits Coronavirus ion channels. Amantadine hydrochloride also has anti-orthopoxvirus and anticancer activity. Amantadine hydrochloride can be used for Parkinson's disease, postoperative cognitive dysfunction (POCD) and COVID-19 research.
Specifications
Specifications
| Chemical Name or Material | Amantadine hydrochloride |
| Physical Form | Solid |
| CAS | 665-66-7 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C10H18ClN |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | 1-Adamantanamine hydrochloride 1-Adamantylamine hydrochloride 1-Aminoadamantane hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (532.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (266.37 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction