missing translation for 'onlineSavingsMsg'
Learn More

C188-9, MedChemExpress

Product Code. 16400917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400917

Brand: Medchem Express HY112288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress C188-9 (TTI-101) is a STAT3 inhibitor, with a Kd of 4.7 nM. C188-9 inhibits G-CSF-induced STAT3 activation and STAT3-dependent gene expression. C188-9 induces apoptosis in AML cell lines and primary samples and inhibits colony formation by primary AML blasts.

C188-9 (TTI-101) is a STAT3 inhibitor, with a Kd of 4.7 nM. C188-9 inhibits G-CSF-induced STAT3 activation and STAT3-dependent gene expression. C188-9 induces apoptosis in AML cell lines and primary samples and inhibits colony formation by primary AML blasts.

Specifications

Chemical Name or Material C188-9
CAS 432001-19-9
Color Pink
Purity Grade Notes Research
Molecular Formula C27H21NO5S
Quantity 50 mg
Synonym TTI-101
Solubility Information DMSO : 25 mg/mL (53.02 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=C3C=CC=CC3=C(O)C(C4=C5C=CC=CC5=CC=C4O)=C2)=O
Molecular Weight (g/mol) 471.52
Formula Weight 471.52
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.