missing translation for 'onlineSavingsMsg'
Learn More

GSK2239633A, MedChemExpress

Product Code. 16495567
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495567 5 mg 5mg
16479486 50 mg 50mg
16460539 1 mg 1mg
16452828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495567 Supplier Medchem Express Supplier No. HY100183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2239633A is a CC-chemokine receptor 4 (CCR4) antagonist, which inhibits the binding of [125I]-TARC to human CCR4 with a pIC50 of 7.96 ± 0.11.

GSK2239633A is a CC-chemokine receptor 4 (CCR4) antagonist, which inhibits the binding of [125I]-TARC to human CCR4 with a pIC50 of 7.96 ± 0.11.

Specifications

Chemical Name or Material GSK2239633A
CAS 1240516-71-5
Color White
Purity Grade Notes Research
Molecular Formula C24H25ClN4O5S2
Quantity 5 mg
Solubility Information DMSO : ≥ 250 mg/mL (455.32 mM)
SMILES CC(C)(O)C(NCC1=CC=CC(CN2N=C(NS(=O)(C3=CC=C(Cl)S3)=O)C4=C2C=CC=C4OC)=C1)=O
Molecular Weight (g/mol) 549.06
Formula Weight 549.06
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.