missing translation for 'onlineSavingsMsg'
Learn More

GSK2200150A, MedChemExpress

Product Code. 16477277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477277 5 mg 5mg
16460867 50 mg 50mg
16434568 10 mg 10mg
16413136 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477277 Supplier Medchem Express Supplier No. HY1120915mg
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK2200150A, identified by high-throughput screening (HTS) campaign, is an anti-tuberculosis (TB) agent.

GSK2200150A, identified by high-throughput screening (HTS) campaign, is an anti-tuberculosis (TB) agent.

Specifications

Chemical Name or Material GSK2200150A
CAS 1443138-53-1
Color White
Purity Grade Notes Research
Molecular Formula C20H23NO3S
Quantity 5 mg
Solubility Information DMSO : 65 mg/mL (181.83 mM; Need ultrasonic)
SMILES N1(CC2=CC=C(OCCO3)C3=C2)CCC4(C(C=CS5)=C5CCO4)CC1
Molecular Weight (g/mol) 357.47
Formula Weight 357.47
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.