missing translation for 'onlineSavingsMsg'
Learn More

GSK2194069, MedChemExpress

Product Code. 16457817
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457817

Brand: Medchem Express HY12325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2194069 is a potent inhibitor of β-ketoyl reductase (KR) of fatty acid synthase (FAS), with an IC50 value of 7.7 nM. GSK2194069 shows specifically inhibitory effect on FAS expressing cancer cells, by acting potent efficacy on acetoacetyl-CoA, NADPH with IC50 or Ki values of 4.8 nM and 5.6 nM, respectively.

GSK2194069 is a potent inhibitor of β-ketoyl reductase (KR) of fatty acid synthase (FAS), with an IC50 value of 7.7 nM. GSK2194069 shows specifically inhibitory effect on FAS expressing cancer cells, by acting potent efficacy on acetoacetyl-CoA, NADPH with IC50 or Ki values of 4.8 nM and 5.6 nM, respectively.

Specifications

Chemical Name or Material GSK2194069
Physical Form Solid
CAS 1332331-08-4
Color Earth Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H24N4O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (233.38 mM)
SMILES O=C(N1C[C@H](CC2=NNC(N2C3=CC=C(C4=CC=C(OC=C5)C5=C4)C=C3)=O)CC1)C6CC6
Molecular Weight (g/mol) 428.48
Formula Weight 428.48
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.