missing translation for 'onlineSavingsMsg'
Learn More

GSK2110183 analog 1 hydrochloride, MedChemExpress

Product Code. 16439487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439487

Brand: Medchem Express HY15966A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2110183 analog 1 hydrochloride is the structural analogue of GSK2110183.

GSK2110183 analog 1 hydrochloride is the structural analogue of GSK2110183.

Specifications

Chemical Name or Material GSK2110183 analog 1 hydrochloride
CAS 2070009-64-0
Color White
Purity Grade Notes Research
Molecular Formula C18H17Cl3F2N4OS
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (207.57 mM; Need ultrasonic) ∣H2O : 25 mg/mL (51.89 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)S1)N[C@@H](CC3=CC=C(F)C(F)=C3)CN.[H]Cl
Molecular Weight (g/mol) 481.77
Formula Weight 481.77
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.