missing translation for 'onlineSavingsMsg'
Learn More

GSK2033, MedChemExpress

Product Code. 16496937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496937 5 mg 5mg
16492996 25 mg 25mg
16484218 10 mg 10mg
16480557 50 mg 50mg
16465166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496937 Supplier Medchem Express Supplier No. HY108688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2033 is a LXR antagonist with pIC50s of 7 and 7.4 for LXRα or LXRβ, respectively.

GSK2033 is a LXR antagonist with pIC50s of 7 and 7.4 for LXRα or LXRβ, respectively.

Specifications

Chemical Name or Material GSK2033
CAS 1221277-90-2
Color White
Purity Grade Notes Research
Molecular Formula C29H28F3NO5S2
Quantity 5 mg
Solubility Information DMSO : 30 mg/mL (50.70 mM; Need ultrasonic and warming)
SMILES O=S(C1=C(C)C=C(C)C=C1C)(N(CC2=CC=C(C3=CC=CC(S(=O)(C)=O)=C3)C=C2)CC4=CC=C(C(F)(F)F)O4)=O
Molecular Weight (g/mol) 591.66
Formula Weight 591.66
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.