missing translation for 'onlineSavingsMsg'
Learn More

GSK1940029, MedChemExpress

Product Code. 16493837
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493837 50 mg 50mg
16467706 100 mg 100mg
16460358 5 mg 5mg
16441859 1 mg 1mg
16427898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493837 Supplier Medchem Express Supplier No. HY19762
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK1940029 (SCD inhibitor 1) is a stearoyl-coa desaturase (SCD) inhibitor extracted from patent WO/2009060053 A1, compound example 16.

GSK1940029 (SCD inhibitor 1) is a stearoyl-coa desaturase (SCD) inhibitor extracted from patent WO/2009060053 A1, compound example 16.

Specifications

Chemical Name or Material GSK1940029
CAS 1150701-66-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H16Cl2N4O2
Quantity 50 mg
Synonym SCD inhibitor 1
Solubility Information DMSO : ≥ 32 mg/mL (81.79 mM)
SMILES ClC(C=CC(CN1N=NC(C(NC2=CC=C(CO)C=C2)=O)=C1C)=C3)=C3Cl
Molecular Weight (g/mol) 391.25
Formula Weight 391.25
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.