missing translation for 'onlineSavingsMsg'
Learn More

GSK1904529A, MedChemExpress

Product Code. p-200100358
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16480377

Brand: Medchem Express HY10524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK1904529A is a potent, selective, orally active, and ATP-competitive inhibitor of insulin-like growth factor-1 receptor (IGF-1R) and insulin receptor (IR), with IC50s of 27 and 25 nM, respectively. GSK1904529A shows poor activity (IC50>1 μM) in 45 other serine/threonine and tyrosine kinases. GSK1904529A exhibits anti-tumor activity.

GSK1904529A is a potent, selective, orally active, and ATP-competitive inhibitor of insulin-like growth factor-1 receptor (IGF-1R) and insulin receptor (IR), with IC50s of 27 and 25 nM, respectively. GSK1904529A shows poor activity (IC50>1 μM) in 45 other serine/threonine and tyrosine kinases. GSK1904529A exhibits anti-tumor activity.

Specifications

Chemical Name or Material GSK1904529A
CAS 1089283-49-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C44H47F2N9O5S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (58.69 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=C(N3C=CC=CC3=N2)C4=NC(NC5=CC(CC)=C(C=C5OC)N6CCC(CC6)N7CCN(CC7)S(=O)(C)=O)=NC=C4)=CC=C1OC)NC8=C(C=CC=C8F)F
Molecular Weight (g/mol) 851.96
Formula Weight 851.96
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.