missing translation for 'onlineSavingsMsg'
Learn More

GSK189254A, MedChemExpress

Product Code. 16458587
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16458587

Brand: Medchem Express HY14111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively.

GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively.

Specifications

Chemical Name or Material GSK189254A
CAS 720690-73-3
Color off White
Purity Grade Notes Research
Molecular Formula C21H25N3O2
Quantity 5 mg
Synonym GSK189254
Solubility Information DMSO : 25 mg/mL (71.14 mM; Need ultrasonic)
SMILES O=C(NC)C(C=N1)=CC=C1OC2=CC(CC3)=C(C=C2)CCN3C4CCC4
Molecular Weight (g/mol) 351.44
Formula Weight 351.44
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.