missing translation for 'onlineSavingsMsg'
Learn More

GSK1838705A, MedChemExpress

Product Code. p-200101160
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465488

Brand: Medchem Express HY13020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK1838705A is a potent and reversible IGF-IR and the insulin receptor inhibitor with IC50s of 2.0 and 1.6 nM, respectively. It also inhibits ALK with an IC50 of 0.5 nM.

GSK1838705A is a potent and reversible IGF-IR and the insulin receptor inhibitor with IC50s of 2.0 and 1.6 nM, respectively. It also inhibits ALK with an IC50 of 0.5 nM.

Specifications

Chemical Name or Material GSK1838705A
CAS 1116235-97-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H29FN8O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (187.77 mM)
SMILES FC1=C(C(NC)=O)C(NC2=C3C(NC=C3)=NC(NC4=CC5=C(CCN5C(CN(C)C)=O)C=C4OC)=N2)=CC=C1
Molecular Weight (g/mol) 532.57
Formula Weight 532.57
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.