missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK1838705A, MedChemExpress
Description
GSK1838705A is a potent and reversible IGF-IR and the insulin receptor inhibitor with IC50s of 2.0 and 1.6 nM, respectively. It also inhibits ALK with an IC50 of 0.5 nM.
Specifications
Specifications
| Chemical Name or Material | GSK1838705A |
| CAS | 1116235-97-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C27H29FN8O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (187.77 mM) |
| SMILES | FC1=C(C(NC)=O)C(NC2=C3C(NC=C3)=NC(NC4=CC5=C(CCN5C(CN(C)C)=O)C=C4OC)=N2)=CC=C1 |
| Molecular Weight (g/mol) | 532.57 |
| Formula Weight | 532.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?