missing translation for 'onlineSavingsMsg'
Learn More

GSK1324726A, MedChemExpress

Product Code. 16438567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438567 5 mg 5mg
16425938 10 mg 10mg
16422087 50 mg 50mg
16406376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16438567 Supplier Medchem Express Supplier No. HY13960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK1324726A is a novel, potent, and selective inhibitor of BET proteins with high affinity to BRD2 (IC50=41 nM), BRD3 (IC50=31 nM), and BRD4 (IC50=22 nM).

GSK1324726A is a novel, potent, and selective inhibitor of BET proteins with high affinity to BRD2 (IC50=41 nM), BRD3 (IC50=31 nM), and BRD4 (IC50=22 nM).

Specifications

Chemical Name or Material GSK1324726A
Physical Form Solid
CAS 1300031-52-0
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H23ClN2O3
For Use With (Application) Cancer-programmed cell death
Synonym I-BET726
Solubility Information DMSO : ≥ 46 mg/mL (105.77 mM)
SMILES ClC1=CC=C(N[C@H]2C3=C(C=CC(C4=CC=C(C(O)=O)C=C4)=C3)N(C(C)=O)[C@@H](C)C2)C=C1
Molecular Weight (g/mol) 434.91
Formula Weight 434.91
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.