missing translation for 'onlineSavingsMsg'
Learn More

GSK1292263, MedChemExpress

Product Code. 16464968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464968 10 mg 10mg
16431217 50 mg 50mg
16427707 5 mg 5mg
16415746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464968 Supplier Medchem Express Supplier No. HY12066
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM).

GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM).

Specifications

Chemical Name or Material GSK1292263
CAS 1032823-75-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H28N4O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 20 mg/mL (43.81 mM)
SMILES CC(C1=NOC(N2CCC(CC2)COC3=CN=C(C=C3)C4=CC=C(C=C4)S(C)(=O)=O)=N1)C
Molecular Weight (g/mol) 456.56
Formula Weight 456.56
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.