missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human KIR2DL3 / CD158b2 Protein, Fc Tag

Product Code. 16437819
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg

Product Code. 16437819

Brand: ACROBiosystems KI3H52571mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Killer cell immunoglobulin-like receptor 2DL3 (KIR2DL3) is also known as CD158 antigen-like family member B2, KIR-023GB, Killer inhibitory receptor cl 2-3, MHC class I NK cell receptor, NKAT2a, NKAT2b, Natural killer-associated transcript 2, p58 natural killer cell receptor clone CL-6, p58.2 MHC class-I-specific NK receptor, CD158b2 and KIR2DL3, which is a single-pass type I membrane protein and belongs to the immunoglobulin superfamily.KIR2DL3 is a receptor on natural killer (NK) cells for HLA-C alleles (HLA-Cw1, HLA-Cw3 and HLA-Cw7). KIR2DL3 can inhibit the activity of NK cells thus preventing cell lysis.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.2 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_056952.2
Molecular Weight (g/mol) 51.2 kDa
Quantity 1 mg
Source Human KIR2DL3, Fc Tag (KI3-H5257) is expressed from human 293 cells (HEK293). It contains AA His 22 - His 245 (Accession # NP_056952.2).
Immunogen His 22 - His 245
Storage Requirements -20°C
Gene Alias KIR2DL3,CD158B2,KIRCL23,NKAT2,KIR-023 GB,NKAT2a,NKAT2b,NKAT-2,CD158 B2,GL183,KIR-023 GB,KIR-K7b,KIR-K7c,KIRCL23,NKAT,p58
Common Name KIR2DL3
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.