missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-J2, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK-J2 is an isomer of GSK-J1 that does not have any specific activity. GSK-J1 is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A.
Specifications
Specifications
| Chemical Name or Material | GSK-J2 |
| Physical Form | Solid |
| CAS | 1394854-52-4 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H23N5O2 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 42 mg/mL (107.84 mM) |
| SMILES | O=C(O)CCNC1=NC(C2=CC=CN=C2)=NC(N3CCC(C=CC=C4)=C4CC3)=C1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction