missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 9311 Hydrochloride

Description
GSK 9311 hydrochloride is a negative control for GSK 6853 (Cat. No. 6198)
Specifications
Specifications
| Chemical Name or Material | N-[2,3-Dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-N-ethyl-2-methoxybenzamide hydrochloride |
| Quantity | 25 mg |
| Molecular Formula | C24H31N5O3.HCl |
| Formula Weight | 474 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?