missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-923295, MedChemExpress
Description
GSK-923295 is a special, allosteric inhibitor of centromere-associated protein-E (CENP-E) kinesin motor ATPase activity, with Ki of 3.2±0.2 nM and 1.6± 0.1 nM for human and canine, respectively.
Specifications
Specifications
| Chemical Name or Material | GSK-923295 |
| CAS | 1088965-37-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H38ClN5O4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 30 mg/mL (50.66 mM; Need ultrasonic) |
| SMILES | O=C(N[C@@H](CC1=CC=C(C2=CN3C(C([C@@H](O)C)=CC=C3)=N2)C=C1)CNC(CN(C)C)=O)C4=CC(Cl)=C(C=C4)OC(C)C |
| Molecular Weight (g/mol) | 592.13 |
| Formula Weight | 592.13 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?