missing translation for 'onlineSavingsMsg'
Learn More

GSK-690693, MedChemExpress

Product Code. 16434806
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434806

Brand: Medchem Express HY10249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK-690693 is an ATP-competitive pan-Akt inhibitor with IC50s of 2 nM, 13 nM, 9 nM for Akt1, Akt2 and Akt3, respectively. GSK-690693 is also an AMPK inhibitor, affects Unc-51-like autophagy activating kinase 1 (ULK1) activity and robustly inhibits STING-dependent IRF3 activation.

GSK-690693 is an ATP-competitive pan-Akt inhibitor with IC50s of 2 nM, 13 nM, 9 nM for Akt1, Akt2 and Akt3, respectively. GSK-690693 is also an AMPK inhibitor, affects Unc-51-like autophagy activating kinase 1 (ULK1) activity and robustly inhibits STING-dependent IRF3 activation.

Specifications

Chemical Name or Material GSK-690693
CAS 937174-76-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H27N7O3
Quantity 100 mg
Solubility Information DMSO : 30 mg/mL (70.51 mM; Need ultrasonic) ∣H2O : 5 mg/mL (11.75 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES CC(O)(C)C#CC1=NC=C(OC[C@@H]2CNCCC2)C3=C1N=C(C4=NON=C4N)N3CC
Molecular Weight (g/mol) 425.48
Formula Weight 425.48
Percent Purity 96.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.