missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-690693, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
GSK-690693 is an ATP-competitive pan-Akt inhibitor with IC50s of 2 nM, 13 nM, 9 nM for Akt1, Akt2 and Akt3, respectively. GSK-690693 is also an AMPK inhibitor, affects Unc-51-like autophagy activating kinase 1 (ULK1) activity and robustly inhibits STING-dependent IRF3 activation.
Specifications
Specifications
| Chemical Name or Material | GSK-690693 |
| CAS | 937174-76-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27N7O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 30 mg/mL (70.51 mM; Need ultrasonic) ∣H2O : 5 mg/mL (11.75 mM; ultrasonic and adjust pH to 5 with HCl) |
| SMILES | CC(O)(C)C#CC1=NC=C(OC[C@@H]2CNCCC2)C3=C1N=C(C4=NON=C4N)N3CC |
| Molecular Weight (g/mol) | 425.48 |
| Formula Weight | 425.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction