missing translation for 'onlineSavingsMsg'
Learn More

GSK 690 Hydrochloride, MedChemExpress

Product Code. 16401127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401127 50 mg 50mg
16477617 5 mg 5mg
16464868 10 mg 10mg
16463266 25 mg 25mg
16405656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16401127 Supplier Medchem Express Supplier No. HY117226A
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK 690 (Hydrochloride) is a reversible inhibitor of lysine specific demethylase 1 (LSD1), with a Kd value of 9 nM and a biochemical IC50 of 37 nM.

GSK 690 (Hydrochloride) is a reversible inhibitor of lysine specific demethylase 1 (LSD1), with a Kd value of 9 nM and a biochemical IC50 of 37 nM.

Specifications

Chemical Name or Material GSK 690 Hydrochloride
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H24ClN3O
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (307.94 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C2=CC(OC[C@H]3CNCC3)=CN=C2C4=CC=C(C)C=C4)C=C1.Cl
Molecular Weight (g/mol) 405.92
Formula Weight 405.92
Percent Purity 99.16%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.