missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 5959
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK 5959 is a potent and selective BRPF1 bromodomain inhibitor (IC50 = 80 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of 35 other bromodomains, including BRPF2/3 and BET family bromodomains. Inhibits BRPF1 interaction with histone H3.3. Cell permeable.
Specifications
Specifications
| Purity | >99% |
| Quantity | 50 mg |
| Product Type | GSK 5959 |
| Molecular Weight (g/mol) | 394.47 |
| Inhibitors | GSK 5959 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction