missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 5959

Product Code. 18709228
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18709228

Brand: Tocris Bioscience™ 5385/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective BRPF1 inhibitor

GSK 5959 is a potent and selective BRPF1 bromodomain inhibitor (IC50 = 80 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of 35 other bromodomains, including BRPF2/3 and BET family bromodomains. Inhibits BRPF1 interaction with histone H3.3. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Purity >99%
Quantity 50 mg
Product Type GSK 5959
Molecular Weight (g/mol) 394.47
Inhibitors GSK 5959

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.