missing translation for 'onlineSavingsMsg'
Learn More

GSK-5498A, MedChemExpress

Product Code. 16427957
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427957 5 mg 5mg
16435238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16427957 Supplier Medchem Express Supplier No. HY12521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK-5498A is a selective small molecule blocker of CARC (IC50, 1 μM); inhibits mediator release from mast cells, and pro-inflammatory cytokine release from T-cells in a variety of species.

GSK-5498A is a selective small molecule blocker of CARC (IC50, 1 μM)

Specifications

Chemical Name or Material GSK-5498A
CAS 1253186-49-0
Color White
Purity Grade Notes Research
Molecular Formula C18H11F6N3O
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (250.44 mM)
SMILES O=C(NC1=NN(CC2=C(F)C=CC=C2C(F)(F)F)C=C1)C3=C(F)C=CC=C3F
Molecular Weight (g/mol) 399.29
Formula Weight 399.29
Percent Purity 97.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.