missing translation for 'onlineSavingsMsg'
Learn More

GSK 4716, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001346
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001346 10 mg 10mg
18025092 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001346 Supplier Tocris Bioscience Supplier No. 3075/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective ERRγ and ERRβ agonist

Selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays selectivity over ERRα and the classical estrogen receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101574-65-6
Molecular Formula C17H18N2O2
Molecular Weight (g/mol) 282.34
MDL Number MFCD00567155
InChI Key IKPPIUNQWSRCOZ-WOJGMQOQSA-N
Synonym e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
PubChem CID 5331325
ChEBI CHEBI:80000
IUPAC Name 4-hydroxy-N'-[(E)-[4-(propan-2-yl)phenyl]methylidene]benzohydrazide
SMILES CC(C)C1=CC=C(\C=N\NC(=O)C2=CC=C(O)C=C2)C=C1

Specifications

Quantity 10 mg
Formula Weight 282.34
Percent Purity >99%
Chemical Name or Material GSK 4716
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.