missing translation for 'onlineSavingsMsg'
Learn More

GSK-4716, MedChemExpress

Product Code. 16477816
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477816 100 mg 100mg
16448048 10 mg 10mg
16423967 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477816 Supplier Medchem Express Supplier No. HY33353
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK-4716 is a selective ERRβ/γ agonist.

GSK-4716 is a selective ERRβ/γ agonist.

Specifications

Chemical Name or Material GSK-4716
CAS 101574-65-6
Color White
Purity Grade Notes Research
Molecular Formula C17H18N2O2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (354.18 mM)
SMILES O=C(N/N=C/C1=CC=C(C(C)C)C=C1)C2=CC=C(O)C=C2
Molecular Weight (g/mol) 282.34
Formula Weight 282.34
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.