missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 429286
GREENER_CHOICE

Product Code. p-5545815
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18064422

Brand: Tocris Bioscience™ 3726/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective Rho-kinase (ROCK) inhibitor

GSK 429286 is a selective Rho-kinase inhibitor (IC50 values are 14, 780 and 1940 nM for ROCK1, RSK and p70S6K respectively). Reverses adrenaline-induced contraction of the rat aortic ring (IC50 = 190 nM) and causes a dose-dependent decrease in mean arterial blood pressure in spontaneous hypertensive rats. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Purity >97%
Gene Alias GSK429286A
Quantity 50 mg
Product Type GSK 429286
Molecular Weight (g/mol) 432.37
Inhibitors GSK 429286

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.