missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 429286

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
GSK 429286 is a selective Rho-kinase inhibitor (IC50 values are 14, 780 and 1940 nM for ROCK1, RSK and p70S6K respectively). Reverses adrenaline-induced contraction of the rat aortic ring (IC50 = 190 nM) and causes a dose-dependent decrease in mean arterial blood pressure in spontaneous hypertensive rats. Orally active.
Specifications
Specifications
| Purity | >97% |
| Gene Alias | GSK429286A |
| Quantity | 50 mg |
| Product Type | GSK 429286 |
| Molecular Weight (g/mol) | 432.37 |
| Inhibitors | GSK 429286 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction