missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 4112

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK 4112 is a selective Rev-Erbα agonist (EC50 = 250 nM). Enhances recruitment of nuclear receptor co-repressor (NCoR) peptide to Rev-Erbα; causes acute suppression of Bmal1 gene transcription. Induces phase shifts in the circadian clock.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | GSK 4112 |
| Purity | >98% |
| Molecular Weight (g/mol) | 396.89 |
| Inhibitors | GSK 4112 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction