missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 4112

Description
GSK 4112 is a selective Rev-Erbα agonist (EC50 = 250 nM). Enhances recruitment of nuclear receptor co-repressor (NCoR) peptide to Rev-Erbα; causes acute suppression of Bmal1 gene transcription. Induces phase shifts in the circadian clock.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | GSK 4112 |
| Purity | >98% |
| Molecular Weight (g/mol) | 396.89 |
| Inhibitors | GSK 4112 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?