missing translation for 'onlineSavingsMsg'
Learn More

GSK 4027, MedChemExpress

Product Code. 16463238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463238 10 mg 10mg
16445967 5 mg 5mg
16439786 50 mg 50mg
16424506 100 mg 100mg
16412686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463238 Supplier Medchem Express Supplier No. HY101027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK 4027 is a chemical probe for the PCAF/GCN5 bromodomain with an pIC50 of 7.4±0.11 for PCAF in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay.

GSK 4027 is a chemical probe for the PCAF/GCN5 bromodomain with an pIC50 of 7.4±0.11 for PCAF in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay.

Specifications

Chemical Name or Material GSK 4027
CAS 2079896-25-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H21BrN4O
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (132.53 mM; Need ultrasonic)
SMILES BrC1=C(N[C@@H]2C[C@H](C3=CC=CC=C3)CN(C)C2)C=NN(C)C1=O
Molecular Weight (g/mol) 377.28
Formula Weight 377.28
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.