missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 beta Antibody (9G11), Novus Biologicals™
Rabbit Monoclonal Antibody
£274.00 - £406.00
Specifications
| Antigen | GSK-3 beta |
|---|---|
| Clone | 9G11 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043769
|
Novus Biologicals
NBP3-26542-50ul |
50 μL |
£274.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043081
|
Novus Biologicals
NBP3-26542-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| GSK-3 beta | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
| A synthesized peptide derived from Human GSK-3 beta [UniProt P49841] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9G11 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2932 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title