Learn More
GSK-3 beta Antibody (272536), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB2506A
Product Code. 18658703
  • £516.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GSK-3 beta
Monoclonal
APC
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
272536
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
E. coli-derived recombinant human GSK-3 beta , Met1-Thr420, Accession # P49841
0.1 mL
Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
2932
Store at 4°C in the dark.
IgG2b