missing translation for 'onlineSavingsMsg'
Learn More

GSK-3 alpha Rabbit anti-Human, Mouse, Rat, Clone: 4I10Q9, Novus Biologicals™

Product Code. 18323066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323066 100 μg 100µL
18387084 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18323066 Supplier Novus Biologicals Supplier No. NBP315639100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GSK-3 alpha Monoclonal antibody specifically detects GSK-3 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK-3 alpha
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 4I10Q9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GSK-3 alpha (P49840).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Hypoxia, Protein Kinase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2931
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.