missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 264220A
GREENER_CHOICE

Product Code. 18024732
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024732 10 mg 10mg
18063942 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024732 Supplier Tocris Bioscience™ Supplier No. 3752/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endothelial lipase and lipoprotein lipase inhibitor

Endothelial lipase and lipoprotein lipase inhibitor (IC50 values are 0.13 and 0.10 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685506-42-7
Molecular Formula C17H21N3O4S

Specifications

Quantity 10 mg
Formula Weight 363.43
Purity >98%
Molecular Weight (g/mol) 363.432
Chemical Name or Material GSK 264220A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.