missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 2606414
GREENER_CHOICE

Product Code. 18776219
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776219

Brand: Tocris Bioscience™ 5107/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PERK inhibitor; orally bioavailable

GSK 2606414 is a potent and selective protein kinase R-like ER kinase (PERK) inhibitor (IC50 = 0.4 nM). Exhibits >1000-fold selectivity for PERK over HR1 and PKR. Inhibits thapsigargin-induced PERK phosphorylation in lung carcinoma A549 cells. Attenuates subcutaneous pancreatic human tumor xenograft growth in mice. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type GSK 2606414
Purity >99%
Molecular Weight (g/mol) 451.44
Inhibitors GSK 2606414

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.